missing translation for 'onlineSavingsMsg'
Learn More

cis-3-Hexen-1-ol, 98%

Product Code. p-3758164 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
This item is not returnable. View return policy

Product Code. 10540381

Brand: Thermo Scientific Acros 120770100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 928-96-1 | C6H12O | 100.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 928-96-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00063217
InChI Key UFLHIIWVXFIJGU-ARJAWSKDSA-N
Synonym z-hex-3-en-1-ol, cis-3-hexen-1-ol, cis-3-hexenol, leaf alcohol, blatteralkohol, z-3-hexenol, z-3-hexen-1-ol, 3-hexenol, 3z-hex-3-en-1-ol, 3-hexen-1-ol, z
PubChem CID 5281167
ChEBI CHEBI:28857
SMILES CC\C=C/CCO

Specifications

Color Colorless to Yellow
Density 0.8480g/mL
Boiling Point 156.0°C to 157.0°C
Flash Point 59°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5CH=CHCH2CH2OH
Refractive Index 1.4380 to 1.4410
Quantity 10 mL
Beilstein 01, I, 229
Merck Index 10, 4592
Specific Gravity 0.848
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and propylene glycol, miscible with oils
IUPAC Name (3Z)-hex-3-en-1-ol
Formula Weight 100.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material cis-3-Hexen-1-ol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • cis-3-Hexen-1-ol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.