missing translation for 'onlineSavingsMsg'
Learn More

cis-3-Hexene, 97%

Product Code. 15440358
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15440358 5 g 5g
15450358 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15440358 Supplier Thermo Scientific Alfa Aesar Supplier No. H53485.06
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 7642-09-3 | C6H12 | 84.16 g/mol

cis-3-Hexene is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-3-Hexene is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7642-09-3
Molecular Weight (g/mol) 84.16
MDL Number MFCD00085300
InChI Key ZQDPJFUHLCOCRG-WAYWQWQTSA-N
Synonym cis-3-hexene, z-3-hexene, 3-hexene, z, z-hex-3-ene, 3z-hex-3-ene, 3-hexene, 3z, unii-50181bz6yt, 3z-3-heptene, 3z-3-hexene #, 3-hexene cis
PubChem CID 643783
IUPAC Name (Z)-hex-3-ene
SMILES CCC=CCC

Specifications

Density 0.681
Boiling Point 66°C to 68°C
Flash Point −25°C (−13°F)
Molecular Formula C6H12
Refractive Index 1.395
Quantity 5 g
UN Number UN3295
Beilstein 1718858
Formula Weight 84.16
Percent Purity 97%
Chemical Name or Material cis-3-Hexene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • cis-3-Hexene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P331-Do NOT induce vomiting.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contain: 3-Hexene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.