missing translation for 'onlineSavingsMsg'
Learn More

cis-3-Nonen-1-ol, 97%

Product Code. 11423448
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423448 25 g 25g
11413448 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11423448 Supplier Thermo Scientific Alfa Aesar Supplier No. A17846.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 10340-23-5 | C9H18O | 142.242 g/mol

Used as flavoring agent for fresh, tropical, melon notes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as flavoring agent for fresh, tropical, melon notes.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10340-23-5
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
MDL Number MFCD00010314
InChI Key IFTBJDZSLBRRMC-SREVYHEPSA-N
Synonym cis-3-nonen-1-ol, z-non-3-en-1-ol, z-3-nonen-1-ol, 3-nonen-1-ol, z, unii-w8g7341qj8, z-3-nonenol, 3z-nonen-1-ol, 3z-3-nonen-1-ol, 3z-non-3-en-1-ol, 3 z ?-?nonenol
PubChem CID 5364631
IUPAC Name (Z)-non-3-en-1-ol
SMILES CCCCCC=CCCO

Specifications

Density 0.847
Boiling Point 107°C to 109°C (20 mmHg)
Flash Point 95°C (203°F)
Refractive Index 1.45
Quantity 25 g
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 142.24
Percent Purity 97%
Chemical Name or Material cis-3-Nonen-1-ol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.