missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Hydroxy-L-proline, 99%

Product Code. 15483627
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15483627 250 mg 250mg
11382159 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15483627 Supplier Thermo Scientific Alfa Aesar Supplier No. H27194.MD
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 618-27-9 | C5H9NO3 | 131.13 g/mol

cis-4-Hydroxy-L-proline (CHP) is a proline analog that inhibits collagen synthesis and has been used as an anticancer compound. CHP blocked myotube formation and expression of sarcomeric myosin heavy chain in C2C12 murine skeletal muscle cells. CHP inhibited proliferation of murine Panc02 pancreatic carcinoma cell line3 and rat pancreatic carcinoma cell line DSL6A. cis-4-Hydroxy-L-proline and other hydrosyl-L-prolines are important intermediates for chiral synthesis of potential drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618-27-9
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.13
MDL Number MFCD00064319,MFCD00064320
InChI Key PMMYEEVYMWASQN-IMJSIDKUSA-N
Synonym cis-4-hydroxy-l-proline, 2s,4s-4-hydroxypyrrolidine-2-carboxylic acid, l-hydroxyproline, l-allohydroxyproline, allo-4-hydroxy-l-proline, cis-4-hydroxyproline, 4s-4-hydroxy-l-proline, l-cis-4-hydroxyproline, l-allo-hydroxyproline, l-proline, 4-hydroxy-, 4s
PubChem CID 440015
ChEBI CHEBI:28397
IUPAC Name (2S,4S)-4-hydroxypyrrolidine-2-carboxylic acid
SMILES O[C@@H]1CN[C@@H](C1)C(O)=O

Specifications

Melting Point ∽250°C (decomposition)
Solubility Information Soluble in water.
Quantity 250 mg
Formula Weight 131.13
Optical Rotation −59° (c=2 in Water)
Percent Purity 99%
Chemical Name or Material cis-4-Hydroxy-L-proline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.