missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Hydroxycyclohexylamine hydrochloride, 95%

Product Code. 15402699
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15402699 1 g 1g
15480087 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15402699 Supplier Thermo Scientific Alfa Aesar Supplier No. H66110.03
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 56239-26-0 | C6H14ClNO | 151.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56239-26-0
Molecular Formula C6H14ClNO
Molecular Weight (g/mol) 151.63
MDL Number MFCD00012566,MFCD07366531,MFCD06410647
InChI Key RKTQEVMZBCBOSB-UHFFFAOYSA-N
Synonym trans-4-aminocyclohexanol hydrochloride, cis-4-aminocyclohexanol hydrochloride, 4-aminocyclohexanol hydrochloride, cis-4-aminocyclohexanol hcl, 4-aminocyclohexan-1-ol hydrochloride, cis-4-amino-cyclohexanol hydrochloride, 4-amino-cyclohexanol hydrochloride, 1r,4r-4-aminocyclohexan-1-ol hydrochloride, cyclohexanol, 4-amino-, hydrochloride, trans-4-aminocyclohexanol hcl
PubChem CID 522619
IUPAC Name 4-aminocyclohexan-1-ol;hydrochloride
SMILES [H+].[Cl-].NC1CCC(O)CC1

Specifications

Melting Point 193°C to 200°C
Quantity 1 g
Beilstein 3909295
Sensitivity Moisture sensitive
Formula Weight 151.63
Percent Purity 95%
Physical Form Solid
Chemical Name or Material cis-4-Hydroxycyclohexylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.