missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Isopropylcyclohexanecarboxylic acid, 97%

Product Code. 15499098
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499098 1 g 1g
15407616 250 mg 250mg
15409108 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15499098 Supplier Thermo Scientific Alfa Aesar Supplier No. H62594.03
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7084-93-7 | C10H18O2 | 170.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7084-93-7
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.25
MDL Number MFCD04004145,MFCD01734696,MFCD19706019
InChI Key YRQKWRUZZCBSIG-UHFFFAOYSA-N
Synonym trans-4-isopropylcyclohexanecarboxylic acid, trans-4-isopropylcyclohexane carboxylic acid, 4-isopropylcyclohexanecarboxylic acid, hexahydrocumic acid, isopropyl-cyclohexanecarboxylic acid, cis-4-isopropylcyclohexanecarboxylic acid, p-isopropylhexahydrobenzoic acid, cyclohexanecarboxylic acid, 4-1-methylethyl, cyclohexanecarboxylic acid, 4-isopropyl, cyclohexanecarboxylic acid, 4-1-methylethyl-, trans
PubChem CID 81526
IUPAC Name 4-propan-2-ylcyclohexane-1-carboxylic acid
SMILES CC(C)C1CCC(CC1)C(O)=O

Specifications

Quantity 1 g
Formula Weight 170.24
Percent Purity 97%
Physical Form Solid
Chemical Name or Material cis-4-Isopropylcyclohexanecarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.