Learn More
cis-4-Octene, 97%
Description
cis-4-octene is used as a chain transfer agent in polymerization reactions, and as a depolymerizing agent in epoxy-functionalized oligomers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 10 g |
| UN Number | UN3295 |
| Solubility Information | Immiscible with water. |
| IUPAC Name | (4Z)-oct-4-ene |
| Formula Weight | 112.22 |
| Percent Purity | 97% |
| Physical Form | Liquid |
| Chemical Name or Material | cis-4-Octene |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.