missing translation for 'onlineSavingsMsg'
Learn More

cis 5-Fluoro-1-[2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-2,4(1H,3H)- pyrimidinedione, TRC

Product Code. 30405281
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30405281 10 mg
30400090 25 mg
30399855 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30405281 Supplier Toronto Research Chemicals Supplier No. TRCF59185010MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145986-11-4
Molecular Formula C8H9FN2O4S
Molecular Weight (g/mol) 248.23
Synonym (2R-cis)-5-Fluoro-1-[2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-2,4(1H,3H)-pyrimidinedione
SMILES O=C1C(F)=CN([C@@H]2CS[C@H](CO)O2)C(N1)=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C8H9FN2O4S/c9-4-1-11(8(14)10-7(4)13)5-3-16-6(2-12)15-5/h1,5-6,12H,2-3H2,(H,10,13,14)/t5-,6+/m0/s1
Chemical Name or Material cis 5-Fluoro-1-[2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-2,4(1H,3H)- pyrimidinedione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.