missing translation for 'onlineSavingsMsg'
Learn More

cis-5-Norbornene-exo-2,3-dicarboxylic anhydride, 95%

Product Code. 15490646
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490646 5 g 5g
15480646 1 g 1g
15400656 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490646 Supplier Thermo Scientific Alfa Aesar Supplier No. H53446.06
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2746-19-2 | C9H8O3 | 164.16 g/mol

cis-5-Norbornene-exo-2,3-dicarboxylic anhydride is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-5-Norbornene-exo-2,3-dicarboxylic anhydride is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2746-19-2
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00630580
InChI Key KNDQHSIWLOJIGP-RNGGSSJXSA-N
Synonym cis-5-norbornene-exo-2,3-dicarboxylic anhydride, 3ar,4r,7s,7as-rel-3a,4,7,7a-tetrahydro-4,7-methanoisobenzofuran-1,3-dione, himic anhydride, unii-n1kqo6559b, norborn-5-ene-2exo,3exo-dicarboxylic acid anhydride, 4,7-methanoisobenzofuran-1,3-dione, 3a,4,7,7a-tetrahydro-, 3aalpha,4beta,7beta,7aalpha-9ci, exo-5-norbornene-2,3-dicarboxylic anhydride, exo-3,6-methylene-1,2,3,6-tetrahydrophthalic anhydride, endicanhydride, 1r,2r,6s,7s-4-oxatricyclo 5.2.1.0 2 ,? dec-8-ene-3,5-dione
PubChem CID 637794
SMILES C1C2C=CC1C3C2C(=O)OC3=O

Specifications

Melting Point 140°C to 145°C
Quantity 5 g
Formula Weight 164.16
Percent Purity 95%
Chemical Name or Material cis-5-Norbornene-exo-2,3-dicarboxylic anhydride
compliance-icons compliance-icons
Product Identifier
  • cis-5-Norbornene-exo-2,3-dicarboxylic anhydride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: Himic anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.