missing translation for 'onlineSavingsMsg'
Learn More

cis-5-Norbornene-exo-2,3-dicarboxylic anhydride, 95%, Thermo Scientific Chemicals

Product Code. 10507824 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10507824 1 g Glass bottle 1g
10776255 5 g Glass bottle 5g
2 options
This item is not returnable. View return policy

Product Code. 10507824

Brand: Thermo Scientific Acros 425740010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2746-19-2
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00630580
InChI Key KNDQHSIWLOJIGP-RNGGSSJXSA-N
Synonym cis-5-norbornene-exo-2,3-dicarboxylic anhydride, 3ar,4r,7s,7as-rel-3a,4,7,7a-tetrahydro-4,7-methanoisobenzofuran-1,3-dione, himic anhydride, unii-n1kqo6559b, norborn-5-ene-2exo,3exo-dicarboxylic acid anhydride, 4,7-methanoisobenzofuran-1,3-dione, 3a,4,7,7a-tetrahydro-, 3aalpha,4beta,7beta,7aalpha-9ci, exo-5-norbornene-2,3-dicarboxylic anhydride, exo-3,6-methylene-1,2,3,6-tetrahydrophthalic anhydride, endicanhydride, 1r,2r,6s,7s-4-oxatricyclo 5.2.1.0 2 ,? dec-8-ene-3,5-dione
PubChem CID 637794
SMILES C1C2C=CC1C3C2C(=O)OC3=O

Specifications

Melting Point 144°C
Quantity 1 g
Packaging Glass bottle
Assay Percent Range 94% min. (GC)
Formula Weight 164.16
Percent Purity 95%
Chemical Name or Material cis-5-Norbornene-exo-2, 3-dicarboxylic anhydride
compliance-icons compliance-icons
Product Identifier
  • cis-5-Norbornene-exo-2,3-dicarboxylic anhydride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.