missing translation for 'onlineSavingsMsg'
Learn More

cis-7,10,13,16-Docosatetraenoic Acid, TRC

Product Code. 30012651
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012651 25 mg 25mg
30012203 100 mg 100mg
30007337 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012651 Supplier Toronto Research Chemicals Supplier No. TRCD49451025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28874-58-0
Molecular Formula C22 H36 O2
Molecular Weight (g/mol) 332.52
Synonym 7Z,10Z,13Z,16Z-Docosatetraenoic Acid, δ7,10,13,16-Docosatetraenoic Acid
IUPAC Name (7Z,10Z,13Z,16Z)-docosa-7,10,13,16-tetraenoic acid
SMILES CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCCCC(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C22H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24/h6-7,9-10,12-13,15-16H,2-5,8,11,14,17-21H2,1H3,(H,23,24)/b7-6-,10-9-,13-12-,16-15-
Formula Weight 332.2715
Chemical Name or Material cis-7,10,13,16-Docosatetraenoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.