missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-Aconitic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Cis-Aconitic Acid |
| CAS | 585-84-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C6 H6 O6 |
| Quantity | 100 mg |
| Synonym | 1-Propene-1,2,3-tricarboxylic acid, (1Z)-, 1-Propene-1,2,3-tricarboxylic acid, (Z)- (8CI), (1Z)-1-Propene-1,2,3-tricarboxylic acid, (Z)-Aconitic acid, NSC 227901, cis-Aconitic acid |
| InChI Formula | InChI=1S/C6H6O6/c7-4(8)1-3(6(11)12)2-5(9)10/h1H,2H2,(H,7,8)(H,9,10)(H,11,12)/b3-1- |
| SMILES | OC(=O)C\C(=C\C(=O)O)\C(=O)O |
| IUPAC Name | (Z)-prop-1-ene-1,2,3-tricarboxylic acid |
| Molecular Weight (g/mol) | 174.11 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?