missing translation for 'onlineSavingsMsg'
Learn More

cis-Aconitic anhydride, 98%

Product Code. 11438616
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438616 5 g 5g
11428616 1 g 1g
11448616 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11438616 Supplier Thermo Scientific Alfa Aesar Supplier No. A16006.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6318-55-4 | C6H4O5 | 156.093 g/mol

cis-Aconitic anhydride is used in the preparation of poly(amino acid)-containing copolymers with gradient pH-sensitive side groups initiated by the amino groups in methoxy poly(ethylene glycol)-block-poly(L-lysine). It is also involved in the preparation of pH-sensitive anti-tumor prodrug nanoparticles by conjugation with D-alfa-tocopheryl polyethylene glycol 1000 succinate (TPGS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-Aconitic anhydride is used in the preparation of poly(amino acid)-containing copolymers with gradient pH-sensitive side groups initiated by the amino groups in methoxy poly(ethylene glycol)-block-poly(L-lysine). It is also involved in the preparation of pH-sensitive anti-tumor prodrug nanoparticles by conjugation with D-alfa-tocopheryl polyethylene glycol 1000 succinate (TPGS).

Notes
Moisture sensitive. Incompatible with strong acids, strong oxidizing agents and strong bases. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6318-55-4
Molecular Formula C6H4O5
Molecular Weight (g/mol) 156.093
MDL Number MFCD00005524
InChI Key GVJRTUUUJYMTNQ-UHFFFAOYSA-N
Synonym cis-aconitic anhydride, aconitic anhydride, cis-aconitic acid anhydride, 3-furanacetic acid, 2,5-dihydro-2,5-dioxo, 2-2,5-dioxo-2,5-dihydrofuran-3-yl acetic acid, 2,5-dioxofuran-3-yl acetic acid, 2-2,5-dioxo-3-furyl acetic acid, 1-propene-1,2,3-tricarboxylic acid, cyclic 1,2-anhydride, 2,5-dioxo-2,5-dihydro-furan-3-yl-acetic acid, z-aconitic anhydride
PubChem CID 65163
IUPAC Name 2-(2,5-dioxofuran-3-yl)acetic acid
SMILES C1=C(C(=O)OC1=O)CC(=O)O

Specifications

Melting Point 74°C to 78°C
Quantity 5 g
UN Number UN3261
Beilstein 127575
Sensitivity Moisture sensitive
Formula Weight 156.1
Percent Purity 98%
Chemical Name or Material cis-Aconitic anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.