missing translation for 'onlineSavingsMsg'
Learn More

cis,cis-Muconic acid, 98%

Product Code. 10053400 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10053400 2.5 g Glass bottle 2.5g
10414725 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10053400 Supplier Thermo Scientific Acros Supplier No. 297760025
 Restricted Item
In Stock
Ajouter au panier
Edge
Ajouter au panier
Request Bulk or Custom Format
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 1119-72-8 | C6H6O4 | 142.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 1119-72-8
Molecular Formula C6H6O4
Molecular Weight (g/mol) 142.11
MDL Number MFCD00085266
InChI Key TXXHDPDFNKHHGW-ZPUQHVIOSA-N
Synonym cis,cis-muconic acid, 2z,4z-hexa-2,4-dienedioic acid, cis,cis-muconate, unii-2g78tuq51g, cis,cis-2,4-hexadienedioic acid, 2,4-hexadienedioic acid, z,z, 2z,4z-hexadienedioic acid, cis,cis-hexadienedioate, z,z-2,4-hexadienedioic acid, 2,4-hexadienedioic acid
PubChem CID 5280518
ChEBI CHEBI:16508
SMILES OC(=O)\C=C\C=C\C(O)=O

Spécification

Melting Point 194°C to 195°C
Color Beige-Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula HO2CCH=CHCH=CHCO2H
Quantity 2.5 g
Merck Index 15, 6385
Solubility Information Solubility in water: freely soluble in boiling water. Other solubilities: sparingly soluble in diethyl ether
IUPAC Name (2E,4E)-hexa-2,4-dienedioic acid
Formula Weight 142.11
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material cis, cis-Muconic acid
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.