missing translation for 'onlineSavingsMsg'
Learn More

cis-Cyclooctene, 95%, stab.

Product Code. 11374549
Change view
Click to view available options
Quantity:
50 mL
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
50mL
Product Code. Quantity unitSize
11374549 50 mL 50mL
11445553 100 mL 100mL
11384549 250 mL 250mL
11455553 500 mL 500mL
4 options
This item is not returnable. View return policy

Product Code. 11374549

Brand: Thermo Scientific Alfa Aesar A13477.AD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 931-87-3 | C8H14 | 110.20 g/mol

cis-Cyclooctene is used as a displaced ligand in chlorobis(cyclooctene)rhodium dimer and chlorobis(cyclooctene)iridium dimer in organometallic chemistry. It acts as a monomer used in synthetic chemistry. It is used to prepare 1-chloro-4-(trichloromethyl) cyclooctane by reaction with carbon tetrachloride using dichlorotris(triphenylphosphine) ruthenium(II) as a catalyst. Further, it is used to study the alkenes chemisorbed on silicon(100) by scanning tunneling microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-Cyclooctene is used as a displaced ligand in chlorobis(cyclooctene)rhodium dimer and chlorobis(cyclooctene)iridium dimer in organometallic chemistry. It acts as a monomer used in synthetic chemistry. It is used to prepare 1-chloro-4-(trichloromethyl) cyclooctane by reaction with carbon tetrachloride using dichlorotris(triphenylphosphine) ruthenium(II) as a catalyst. Further, it is used to study the alkenes chemisorbed on silicon(100) by scanning tunneling microscopy.

Solubility
Miscible with alcohol and ether. Immiscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 931-87-3
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number MFCD00001753
InChI Key URYYVOIYTNXXBN-UPHRSURJSA-N
Synonym cis-cyclooctene, z-cyclooctene, cyclooctene, z, cyclooctene, 1z, cyclooct-1-ene, e-cyclooctene, 1-cyclooctene #, 1z-cyclooctene, 7z-cyclooctene, ar,e-cyclooctene
PubChem CID 638079
SMILES C1CCC\C=C/CC1

Specifications

Melting Point -16°C
Density 0.848
Boiling Point 145°C to 146°C
Flash Point 25°C (77°F)
Odor Recognizable
Refractive Index 1.47
Quantity 50 mL
UN Number UN3295
Beilstein 1280166
Sensitivity Air Sensitive
Solubility Information Miscible with alcohol and ether. Immiscible with water.
IUPAC Name (Z)-cyclooctene
Formula Weight 110.2
Percent Purity 95%
Chemical Name or Material cis-Cyclooctene, stabilized
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.