missing translation for 'onlineSavingsMsg'
Learn More

cis-Decahydro-2a,4a,6a,8a-tetraazacyclopent[fg]acenaphthylene, Thermo Scientific Chemicals

Product Code. 11356939
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11356939 1 g 1g
15427625 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11356939 Supplier Thermo Scientific Alfa Aesar Supplier No. H26815.03
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 79236-92-3 | C10H18N4 | 194.282 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79236-92-3
Molecular Formula C10H18N4
Molecular Weight (g/mol) 194.282
MDL Number MFCD09263319
InChI Key YSPZOYMEWUTYDA-UHFFFAOYSA-N
Synonym cis-glyoxal-cyclen, 2a,4a,6a,8a-decahydrotetraazacyclopent fg acenaphthylene, cis-decahydro-2a,4a,6a,8a-tetraazacyclopent fg acenaphthylene, cyclen cis-glyoxal derivative, 1,1':3,3'-diethano-2,2'-biimidazolidine, decahydro-2a,4a,6a,8a-tetraazacyclopenta fg acenaphthylene, cis-decahydro-2a,4a,6a,8a-tetraazacyclopent fg acena, 1,4,7,10-tetraazatetracyclo-5,5,2,04,13,010,14 tetradecane, 1,4,7,10-tetraazatetracyclo-5,5,2,04.13,010.14 tetradecane, 4aalpha,8abeta-1,8:4,5-diethanodecahydro-1,4,5,8-tetraazanaphthalene
PubChem CID 9794169
SMILES C1CN2CCN3CCN4C3C2N1CC4

Specifications

Melting Point 29°C to 31°C
Quantity 1 g
Sensitivity Hygroscopic
Formula Weight 194.28
Chemical Name or Material cis-Decahydro-2a,4a,6a,8a-tetraazacyclopent[fg]acenaphthylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.