missing translation for 'onlineSavingsMsg'
Learn More

cis-Dichlorobis(2,2'-bipyridine)ruthenium(II) dihydrate, 99%

Product Code. 10552422 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10552422 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10552422 Supplier Thermo Scientific Acros Supplier No. 363300010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15746-57-3 | C20H16Cl2N4Ru·2H2O | 520.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 15746-57-3
Color Black
Elemental Analysis 98.5% min. (ex Ru)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C20H16Cl2N4Ru·2H2O
MDL Number MFCD00015504
Quantity 1 g
Synonym cis-dichlorobis 2,2'-bipyridine ruthenium ii dihydrate, 99, cis-dichlorobis 2,2'-bipyridine-ruthenium ii dihydrate
Solubility Information Solubility in water: soluble
InChI Key NHKTUSUPCAKVHT-UHFFFAOYSA-L
SMILES C1=CC=NC(=C1)C2=CC=CC=N2.C1=CC=NC(=C1)C2=CC=CC=N2.O.O.[Cl-].[Cl-].[Ru+2]
IUPAC Name 2-pyridin-2-ylpyridine;ruthenium(2+);dichloride;dihydrate
Molecular Weight (g/mol) 520.38
PubChem CID 71463877
Formula Weight 520.38
Percent Purity 99%
Physical Form Crystals or Powder
Chemical Name or Material cis-Dichlorobis(2, 2'-bipyridine)ruthenium(II) dihydrate
Show More Show Less
compliance-icons
Product Identifier
  • cis-Dichlorobis(2,2'-bipyridine)ruthenium(II) dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.