missing translation for 'onlineSavingsMsg'
Learn More

cis-Dichlorobis(triethylphosphine)platinum(II)

Product Code. 15486295
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15486295 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15486295 Supplier Thermo Scientific Alfa Aesar Supplier No. 039284.ME
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 15692-07-6 | C12H30Cl2P2Pt | 502.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 15692-07-6
Melting Point 139°C to 142°C
Molecular Formula C12H30Cl2P2Pt
MDL Number MFCD00058821
Quantity 500 mg
Sensitivity Hygroscopic
Synonym cis-dichlorobis triethylphosphine platinum ii, et3p 2ptcl2, trans-dichlorobis triethylphosphine platinum ii, cis-bis triethylphosphine platinum dichloride, bis triethylphosphine platinum dichloride, cis-bis triethylphosphine platinum cento chloride, dichlorobis triethylphosphine platinum ii, cis-bis triethylphosphine platinum ii chloride
InChI Key WPWLTKRUFHHDLP-UHFFFAOYSA-L
SMILES [Cl-].[Cl-].[Pt++].CCP(CC)CC.CCP(CC)CC
IUPAC Name dichloroplatinum;triethylphosphane
Molecular Weight (g/mol) 502.30
PubChem CID 11027520
Formula Weight 502.32
Physical Form Powder
Chemical Name or Material cis-Dichlorobis(triethylphosphine)platinum(II)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.