missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-Dichlorodiamineplatinum(II), 99.99%, (trace metal basis)
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
Specifications
Specifications
| Melting Point | 277.0°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 5 g |
| Assay Percent Range | 64.0 to 66% (Pt) |
| Linear Formula | Pt(NH3)2Cl2 |
| Merck Index | 15, 2316 |
| Solubility Information | Solubility in water: 0.25g/100mL (25°C) |
| Formula Weight | 300.05 |
| Percent Purity | 99.99% |
| Show More |
Safety and Handling
Product Identifier
- cis-Dichlorodiamineplatinum(II)
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H350-May cause cancer.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H300-Fatal if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction