missing translation for 'onlineSavingsMsg'
Learn More

cis-Etoposide, TRC

Product Code. 30010495
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
100 mg
missing translation for 'unitSize':
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30010495 10 mg 10mg
30009183 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30010495 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCE93374010MG
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100007-56-5
Molecular Formula C29 H32 O13
Molecular Weight (g/mol) 588.56
Synonym Furo[3',4':6,7]naphtho[2,3-d]-1,3-dioxol-6(5aH)-one, 9-[[4,6-O-(1R)-ethylidene-β-D-glucopyranosyl]oxy]-5,8,8a,9-tetrahydro-5-(4-hydroxy-3,5-dimethoxyphenyl)-, (5R,5aS,8aR,9S)-, Furo[3',4':6,7]naphtho[2,3-d]-1,3-dioxol-6(5aH)-one, 9-[(4,6-O-ethylidene-β-D-glucopyranosyl)oxy]-5,8,8a,9-tetrahydro-5-(4-hydroxy-3,5-dimethoxyphenyl)-, [5R-[5α,5aα,8aα,9β(R*)]]-, (5R,5aS,8aR,9S)-9-[[4,6-O-(1R)-Ethylidene-β-D-glucopyranosyl]oxy]-5,8,8a,9-tetrahydro-5-(4-hydroxy-3,5-dimethoxyphenyl)furo[3',4':6,7]naphtho[2,3-d]-1,3-dioxol-6(5aH)-one, Pyrano[3,2-d]-1,3-dioxin, furo[3',4':6,7]naphtho[2,3-d]-1,3-dioxol-6(5aH)-one deriv., Picro ethylidene lignan P, Picroetoposide, Pptoxin IV, cis-Etoposide, (5R,5aS,8aR,9S)-9-[[4,6-O-[(1R)-ethane-1,1-diyl]-β-D-glucopyranosyl]oxy]-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,8,8a,9-tetrahydro[2]benzofuro[5,6-f][1,3]benzodioxol-6(5aH)-one
IUPAC Name (5S,5aR,8aS,9R)-5-[[(2R,4aR,6R,7R,8R,8aS)-7,8-dihydroxy-2-methyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-6-yl]oxy]-9-(4-hydroxy-3,5-dimethoxyphenyl)-5a,6,8a,9-tetrahydro-5H-[2]benzofuro[5,6-f][1,3]benzodioxol-8-one
SMILES COc1cc(cc(OC)c1O)[C@H]2[C@H]3[C@H](COC3=O)[C@H](O[C@@H]4O[C@@H]5CO[C@@H](C)O[C@H]5[C@H](O)[C@H]4O)c6cc7OCOc7cc26

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C29H32O13/c1-11-36-9-20-27(40-11)24(31)25(32)29(41-20)42-26-14-7-17-16(38-10-39-17)6-13(14)21(22-15(26)8-37-28(22)33)12-4-18(34-2)23(30)19(5-12)35-3/h4-7,11,15,20-22,24-27,29-32H,8-10H2,1-3H3/t11-,15+,20-,21-,22-,24-,25-,26-,27-,29+/m1/s1
Formula Weight 588.1843
Percent Purity >95%
Chemical Name or Material cis-Etoposide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.