missing translation for 'onlineSavingsMsg'
Learn More

cis-Heptachlor Epoxide, TRC

Product Code. 30404473
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30404473 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30404473 Supplier Toronto Research Chemicals Supplier No. TRCH2657001MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Heptachlor-exo-epoxide (cis-, isomer B)
CAS 1024-57-3
Molecular Formula C10 H5 Cl7 O
Quantity 1 mg
Synonym (1aR,1bS,2R,5S,5aR,6S,6aR)-rel-2,3,4,5,6,7,7-Heptachloro-1a,1b,5,5a,6,6a-hexahydro-2,5-methano-2H-indeno[1,2-b]oxirene, (1aα,1bβ,2α,5α,5aβ,6β,6aα)-2,3,4,5,6,7,7-Heptachloro-1a,1b,5,5a,6,6a-hexahydro-2,5-methano-2H-indeno[1,2-b]oxirene, 1,4,5,6,7,8,8-Heptachloro-2,3-epoxy-3a,4,7,7a-tetrahydro-4,7-methanoindan, (±)-cis-Heptachlor epoxide, ENT 25584, Epoxyheptachlor, GPKh epoxide, HCE, HE, Hepox, Heptachlor cis-epoxide, Heptachlor epoxide, Heptachlor epoxide B, Heptachlor exo-epoxide, Heptepoxide, Velsicol 53CS17, cis-Heptachlor epoxide
InChI Formula InChI=1S/C10H5Cl7O/c11-3-1-2(4-5(3)18-4)9(15)7(13)6(12)8(1,14)10(9,16)17/h1-5H/t1-,2+,3+,4-,5+,8+,9-/m0/s1
SMILES Cl[C@H]1[C@H]2O[C@H]2[C@H]3[C@@H]1[C@@]4(Cl)C(=C(Cl)[C@]3(Cl)C4(Cl)Cl)Cl
Molecular Weight (g/mol) 389.32
Formula Weight 385.816
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.