missing translation for 'onlineSavingsMsg'
Learn More

cis-Heptachlor Epoxide, TRC

Product Code. 30400292
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30400292 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400292 Supplier Toronto Research Chemicals Supplier No. TRCH2657005MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Heptachlor-exo-epoxide (cis-, isomer B)
CAS 1024-57-3
Molecular Formula C10 H5 Cl7 O
Quantity 5 mg
Synonym (1aR,1bS,2R,5S,5aR,6S,6aR)-rel-2,3,4,5,6,7,7-Heptachloro-1a,1b,5,5a,6,6a-hexahydro-2,5-methano-2H-indeno[1,2-b]oxirene, (1aα,1bβ,2α,5α,5aβ,6β,6aα)-2,3,4,5,6,7,7-Heptachloro-1a,1b,5,5a,6,6a-hexahydro-2,5-methano-2H-indeno[1,2-b]oxirene, 1,4,5,6,7,8,8-Heptachloro-2,3-epoxy-3a,4,7,7a-tetrahydro-4,7-methanoindan, (±)-cis-Heptachlor epoxide, ENT 25584, Epoxyheptachlor, GPKh epoxide, HCE, HE, Hepox, Heptachlor cis-epoxide, Heptachlor epoxide, Heptachlor epoxide B, Heptachlor exo-epoxide, Heptepoxide, Velsicol 53CS17, cis-Heptachlor epoxide
InChI Formula InChI=1S/C10H5Cl7O/c11-3-1-2(4-5(3)18-4)9(15)7(13)6(12)8(1,14)10(9,16)17/h1-5H/t1-,2+,3+,4-,5+,8+,9-/m0/s1
SMILES Cl[C@H]1[C@H]2O[C@H]2[C@H]3[C@@H]1[C@@]4(Cl)C(=C(Cl)[C@]3(Cl)C4(Cl)Cl)Cl
Molecular Weight (g/mol) 389.32
Formula Weight 385.816
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.