missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ cis-Stilbeneboronic acid pinacol ester, 99%

Product Code. 11395345
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11395345 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11395345 Supplier Alfa Aesar™ Supplier No. L19576.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 264144-59-4
Molecular Formula C20H23BO2
Molecular Weight (g/mol) 306.21
MDL Number MFCD03788734,MFCD03788734
InChI Key HGZKORGQZNCVQC-UHFFFAOYSA-N
Synonym 2-1,2-diphenylethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 53399103
IUPAC Name 2-(1,2-diphenylethenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C(=CC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 92°C to 94°C
Quantity 250 mg
Beilstein 9269561
Formula Weight 306.22
Percent Purity 99%
Chemical Name or Material cis-Stilbeneboronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.