missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cisapride monohydrate

Product Code. 17201156 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
17201156 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 17201156

Brand: Thermo Scientific Chemicals 465900010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 260779-88-2 | C23H29ClFN3O4·H2O, C23H31ClFN3O5 | 483.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 260779-88-2
Quantity 1 g
Molecular Formula C23H31ClFN3O5
Formula Weight 483.97
InChI Key QBYYXIDJOFZORM-UHFFFAOYNA-N
SMILES O.COC1CN(CCCOC2=CC=C(F)C=C2)CCC1NC(=O)C1=CC(Cl)=C(N)C=C1OC
IUPAC Name 4-amino-5-chloro-N-{1-[3-(4-fluorophenoxy)propyl]-3-methoxypiperidin-4-yl}-2-methoxybenzamide hydrate
Molecular Weight (g/mol) 483.97
Chemical Name or Material Cisapride monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.