missing translation for 'onlineSavingsMsg'
Learn More

Cisatracurium-20-methyl Dibenzenesulfonate, TRC

Product Code. 30352658
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30352658 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30352658 Supplier Toronto Research Chemicals Supplier No. TRCC496705100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cisatracurium-20-methyl Dibenzenesulfonate
CAS 1193104-85-6
Molecular Formula C54 H74 N2 O12 . 2 C6 H5 O3 S
Quantity 100 mg
Synonym (1R,1'R,2R,2'R)-2,2'-((Hexane-1,6-diylbis(oxy))bis(3-oxopropane-3,1-diyl))bis(1-(3,4-dimethoxybenzyl)-6,7-dimethoxy-2-methyl-1,2,3,4-tetrahydroisoquinolin-2-ium) Henzenesulfonate, (1R,1'R,2R,2'R)-2,2'-[1,6-Hexanediylbis[oxy(3-oxo-3,1-propanediyl)]]bis[1-[(3,4-dimethoxyphenyl)methyl]-1,2,3,4-tetrahydro-6,7-dimethoxy-2-methylisoquinolinium Benzenesulfonate (1:2)
InChI Formula InChI=1S/C54H74N2O12.2C6H6O3S/c1-55(23-19-39-33-49(63-7)51(65-9)35-41(39)43(55)29-37-15-17-45(59-3)47(31-37)61-5)25-21-53(57)67-27-13-11-12-14-28-68-54(58)22-26-56(2)24-20-40-34-50(64-8)52(66-10)36-42(40)44(56)30-38-16-18-46(60-4)48(32-38)62-6;2*7-10(8,9)6-4-2-1-3-5-6/h15-18,31-36,43-44H,11-14,19-30H2,1-10H3;2*1-5H,(H,7,8,9)/q+2;;/p-2/t43-,44-,55-,56-;;/m1../s1
SMILES COc1ccc(C[C@@H]2c3cc(OC)c(OC)cc3CC[N@+]2(C)CCC(=O)OCCCCCCOC(=O)CC[N@@+]4(C)CCc5cc(OC)c(OC)cc5[C@H]4Cc6ccc(OC)c(OC)c6)cc1OC.[O-]S(=O)(=O)c7ccccc7.[O-]S(=O)(=O)c8ccccc8
IUPAC Name benzenesulfonate;6-[3-[(1R,2R)-1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-2-methyl-3,4-dihydro-1H-isoquinolin-2-ium-2-yl]propanoyloxy]hexyl 3-[(1R,2R)-1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-2-methyl-3,4-dihydro-1H-isoquinolin-2-ium-2-yl]propanoate
Molecular Weight (g/mol) 1257.506
Formula Weight 1256.516
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.