missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Citalopram hydrobromide

Product Code. 30001252
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001252 10 mg 10mg
18064492 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001252 Supplier R&D Systems™ Supplier No. 1427/10
  • ONLINE EXCLUSIVE PRICE
    225.08 €
    202.65 € / 10mg
    Save 22.43 €
    9% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Highly potent and selective 5-HT uptake inhibitor

Citalopram hydrobromide is a highly selective and potent 5-HT uptake inhibitor with no effect on noradrenalin or dopamine uptake (IC50 values are 1.8, 8800 and 41000 nM respectively). Has negligible activity at a wide range of receptors.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59729-32-7
Molecular Formula C20H22BrFN2O

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 405.31
Purity >99%
Molecular Weight (g/mol) 405.311
Chemical Name or Material Citalopram hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.