missing translation for 'onlineSavingsMsg'
Learn More

Citicoline, 98%

Product Code. 15572323 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15572323 1 g 1g
15582323 5 g 5g
15592323 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15572323 Supplier Thermo Scientific Acros Supplier No. 457780010
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 987-78-0 | C14H26N4O11P2 | 488.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 987-78-0
Molecular Formula C14H26N4O11P2
Molecular Weight (g/mol) 488.33
MDL Number MFCD00868097
InChI Key RZZPDXZPRHQOCG-OJAKKHQRSA-N
Synonym cdp-choline, cidifos, citicoline, cytidine diphosphate choline, cdp-choline 1-, 5'-o-2-trimethylazaniumyl ethoxy phosphinato oxy phosphinato cytidine
PubChem CID 25202509
ChEBI CHEBI:58779
IUPAC Name [[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] 2-(trimethylazaniumyl)ethyl phosphate
SMILES C[N+](C)(C)CCOP([O-])(=O)OP(O)(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=CC(N)=NC1=O

Specifications

Melting Point 240.0°C to 242.0°C
Loss on Drying 5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 488.32
Percent Purity 98%
Chemical Name or Material Citicoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.