missing translation for 'onlineSavingsMsg'
Learn More

Citicoline, MedChemExpress

Product Code. 16478436
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478436 100 mg 100mg
16472206 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16478436 Supplier Medchem Express Supplier No. HYB0739
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Citicoline (Cytidine diphosphate-choline) is an intermediate in the synthesis of phosphatidylcholine, a component of cell membranes. Citicoline exerts neuroprotective effects.

Citicoline (Cytidine diphosphate-choline) is an intermediate in the synthesis of phosphatidylcholine, a component of cell membranes. Citicoline exerts neuroprotective effects.

Specifications

Chemical Name or Material Citicoline
CAS 987-78-0
Color White
Purity Grade Notes Research
Molecular Formula C14H26N4O11P2
Quantity 100 mg
Synonym Cytidine diphosphate-choline CDP-Choline Cytidine 5'-diphosphocholine
Solubility Information H2O : 50 mg/mL (102.39 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (2.05 mM; ultrasonic and warming and heat to 80°C)
SMILES O[C@@H]([C@H]([C@H](N1C(N=C(C=C1)N)=O)O2)O)[C@H]2COP(OCC[N+](C)(C)C)(OP([O-])(O)=O)=O
Molecular Weight (g/mol) 488.32
Formula Weight 488.32
Percent Purity 99.51%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.