missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citicoline sodium, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
Citicoline sodium salt is an intermediate in the synthesis of phosphatidylcholine which is a component of cell membranes and also exerts neuroprotective effects.
Specifications
Specifications
| Chemical Name or Material | Citicoline sodium |
| CAS | 33818-15-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C14H25N4NaO11P2 |
| Quantity | 500 mg |
| Synonym | Cytidine diphosphate-choline sodiumCDP-Choline sodiumCytidine 5'-diphosphocholine sodium |
| Solubility Information | H2O : ≥ 100 mg/mL (195.96 mM) ∣DMSO : 1 mg/mL (1.96 mM; ultrasonic and warming and heat to 80°C) |
| SMILES | O[C@H]1[C@H](N(C=CC(N)=N2)C2=O)O[C@H](COP(OP(OCC[N+](C)(C)C)([O-])=O)(O[Na])=O)[C@H]1O |
| Molecular Weight (g/mol) | 510.31 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction