| CAS |
33818-15-4
|
| Formula Weight |
510.0893 |
| InChI Formula |
InChI=1S/C14H26N4O11P2.Na/c1-18(2,3)6-7-26-30(22,23)29-31(24,25)27-8-9-11(19)12(20)13(28-9)17-5-4-10(15)16-14(17)21;/h4-5,9,11-13,19-20H,6-8H2,1-3H3,(H3-,15,16,21,22,23,24,25);/q;+1/p-1/t9-,11-,12-,13-;/m1./s1 |
| IUPAC Name |
sodium;[[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] 2-(trimethylazaniumyl)ethyl phosphate |
| Molecular Formula |
C14 H25 N4 O11 P2 . Na |
| Molecular Weight (g/mol) |
510.31 |
| Percent Purity |
>95% |
| SMILES |
[Na+].C[N+](C)(C)CCOP(=O)([O-])OP(=O)([O-])OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=CC(=NC2=O)N |
| Synonym |
Choline, hydroxide, monoester with cytidine 5'-(trihydrogen pyrophosphate), inner salt, monosodium salt (8CI), Cytidine 5'-(trihydrogen diphosphate), P'-[2-(trimethylammonio)ethyl] ester, hydroxide, inner salt, monosodium salt, Cytidine 5'-(trihydrogen diphosphate), P'-[2-(trimethylammonio)ethyl] ester, inner salt, monosodium salt (9CI), Acticolin, Brassel, Cebroton, CerAxon, Cidifos, Citicoline sodium, Flussorex, Gerolin, IP 302 sodium, Logan, Neurotron, Sinkron, Strolin |
| Chemical Name or Material |
Citicoline sodium |
| Grade |
HPLC |
|
Show More
Show Less
|
|