missing translation for 'onlineSavingsMsg'
Learn More

Citraconic Acid, TRC

Product Code. 30363693
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.50g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30363693 500 mg 500mg
30355404 2.5 g 2.50g
30353447 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30363693 Supplier Toronto Research Chemicals Supplier No. TRCC520518500MG
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 498-23-7
Molecular Formula C5H6O4
Molecular Weight (g/mol) 130.09
Synonym (2Z)-2-Methyl-2-butenedioic Acid, (Z)-2-Methyl-2-butenedioic Acid, 2-Methylmaleic Acid, Methylmaleic Acid
IUPAC Name (Z)-2-methylbut-2-enedioic acid
SMILES C\C(=C\C(=O)O)\C(=O)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C5H6O4/c1-3(5(8)9)2-4(6)7/h2H,1H3,(H,6,7)(H,8,9)/b3-2-
Formula Weight 130.0266
Chemical Name or Material Citraconic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.