missing translation for 'onlineSavingsMsg'
Learn More

Citraconic anhydride, TRC

Product Code. 30364008
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Unit Size:
1g
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30364008 500 mg 500mg
30362793 2.5 g 2.5g
30365531 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30364008 Supplier Toronto Research Chemicals Supplier No. TRCC520513500MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-02-4
Molecular Formula C5H4O3
Molecular Weight (g/mol) 112.08
Synonym Indobufen Impurity 19
IUPAC Name 3-methylfuran-2,5-dione
SMILES CC1=CC(=O)OC1=O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C5H4O3/c1-3-2-4(6)8-5(3)7/h2H,1H3
Formula Weight 112.016
Chemical Name or Material Citraconic anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.