missing translation for 'onlineSavingsMsg'
Learn More

Citral, 95%, mixture of cis and trans

Product Code. 10498820 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10498820 100 mL Glass bottle 100mL
10010760 5 mL Glass bottle 5mL
10182850 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10498820 Supplier Thermo Scientific Acros Supplier No. 110441000
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5392-40-5 | C10H16O | 152.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5392-40-5
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00006997
InChI Key WTEVQBCEXWBHNA-JXMROGBWSA-N
Synonym citral, geranial, trans-citral, 3,7-dimethylocta-2,6-dienal, geranialdehyde, e-citral, citral a, 2e-3,7-dimethylocta-2,6-dienal, alpha-citral, lemsyn gb
PubChem CID 638011
ChEBI CHEBI:16980
IUPAC Name (2E)-3,7-dimethylocta-2,6-dienal
SMILES CC(C)=CCC\C(C)=C\C=O

Specifications

Melting Point -10°C
Color Yellow
Density 0.8890g/mL
Boiling Point 229°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C=CHCH2CH2C(CH3)=CHCHO
Refractive Index 1.4860 to 1.49
Quantity 100 mL
Merck Index 15, 2321
Specific Gravity 0.889
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in 5 volumes of 60% alcohol,soluble in al,benzyl benzoate,diethyl phthalate,glycerol,prop,mineral oil,fixed oils
Formula Weight 152.24
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Citral
Show More Show Less
compliance-icons
Product Identifier
  • Citral, mixture of cis and trans
Signal Word
  • Warning
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.