missing translation for 'onlineSavingsMsg'
Learn More

Citral, cis + trans, 95%

Product Code. 11483227
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
Unit Size:
100g
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483227 500 g 500g
11354549 50 g 50g
11473227 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11483227 Supplier Thermo Scientific Alfa Aesar Supplier No. A16169.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5392-40-5 | C10H16O | 152.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5392-40-5
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00006997
InChI Key WTEVQBCEXWBHNA-JXMROGBWSA-N
Synonym citral, geranial, trans-citral, 3,7-dimethylocta-2,6-dienal, geranialdehyde, e-citral, citral a, 2e-3,7-dimethylocta-2,6-dienal, alpha-citral, lemsyn gb
PubChem CID 638011
ChEBI CHEBI:16980
IUPAC Name (2E)-3,7-dimethylocta-2,6-dienal
SMILES CC(C)=CCC\C(C)=C\C=O

Specifications

Melting Point <-10°C
Density 0.888
Boiling Point 228°C to 229°C
Flash Point 98°C (208°F)
Refractive Index 1.488
Quantity 500 g
Beilstein 1721871
Merck Index 14,2322
Formula Weight 152.24
Percent Purity 95%
Chemical Name or Material Citral, cis + trans
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.