missing translation for 'onlineSavingsMsg'
Learn More

Citreoviridin (80%, Contain Isocitreoviridin), TRC

Product Code. 30135285
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135285 5 mg 5mg
30133227 2.5 mg 2.50mg
30005253 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135285 Supplier Toronto Research Chemicals Supplier No. TRCC5205605MG
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25425-12-1
Molecular Formula C23H30O6
Molecular Weight (g/mol) 402.48
Synonym Citreoviridin A, Citreoviridine, Citreoviridine A, 5-Hydroxy-3-methoxy-4,12-dimethyl-13-(tetrahydro-3,4-dihydroxy-2,4,5-trimethyl-2-furyl)-2,4,6,8,10,12-tridecahexaenoic Acid, δ-Lactone, [2S-[2α(1E,3E,5E,7E),3β,4α,5α]]-4-Methoxy-5-methyl-6-[7-methyl-8-(tetrahydro-3,4-dihydroxy-2,4,5-trimethyl-2-furanyl)-1,3,5,7-octatetraenyl]-2H-pyran-2-one, 4-Methoxy-5-methyl-6-[7-methyl-8-(tetrahydro-3,4-dihydroxy-2,4,5-trimethyl-2-furyl)-1,3,5,7-octatetraenyl]-2H-pyran-2-one, 2,5-Anhydro-1,6-dideoxy-2-C-[(1E,3E,5E,7E)-8-(4-methoxy-5-methyl-2-oxo-2H-pyran-6-yl)-2-methyl-1,3,5,7-octatetraenyl]-4-C-methyl-D-Iditol, 6-((1E,3E,5E,7E)-8-((2S,3R,4R,5R)-3,4-Dihydroxy-2,4,5-trimethyltetrahydrofuran-2-yl)-7-methylocta-1,3,5,7-tetraen-1-yl)-4-methoxy-5-methyl-2H-pyran-2-one
IUPAC Name 6-[(1E,3E,5E,7E)-8-[(2S,3R,4R,5R)-3,4-dihydroxy-2,4,5-trimethyloxolan-2-yl]-7-methylocta-1,3,5,7-tetraenyl]-4-methoxy-5-methylpyran-2-one
SMILES COC1=CC(=O)OC(=C1C)\C=C\C=C\C=C\C(=C\[C@]2(C)O[C@H](C)[C@](C)(O)[C@H]2O)\C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C23H30O6/c1-15(14-22(4)21(25)23(5,26)17(3)29-22)11-9-7-8-10-12-18-16(2)19(27-6)13-20(24)28-18/h7-14,17,21,25-26H,1-6H3/b8-7+,11-9+,12-10+,15-14+/t17-,21+,22+,23+/m1/s1
Formula Weight 402.2042
Percent Purity >95%
Chemical Name or Material Citreoviridin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.