missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid, 99+%, Technical, Anhydrous, SLR, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical

Product Code. 10599180 Shop All Fisher Chemical Products
Click to view available options
:
500g, Plastic powder jar
This item is not returnable. View return policy

Product Code. 10599180

Brand: Fisher Chemical C/6240/53

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

pH 1.7
CAS 77-92-9
Melting Point 153°C
Color White
Molecular Formula C6H8O7
MDL Number 11669
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
Molecular Weight (g/mol) 192.12
Formula Weight 192.13g/mol
Quantity 500 g
Packaging Plastic powder jar
Physical Form Solid
Show More Show Less
compliance-icons
Product Identifier
  • Citric acid anhydrous
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.