missing translation for 'onlineSavingsMsg'
Learn More

Citric acid, Anhydrous, 99.5 to 100.5% (Dry Basis)

Product Code. p-7082808 Shop All Thermo Scientific Products
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
500g
This item is not returnable. View return policy

Product Code. 15885108

Brand: Thermo Scientific J13729.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 77-92-9 | C6H8O7 | 192.123 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-92-9
Molecular Formula C6H8O7
Molecular Weight (g/mol) 192.123
MDL Number MFCD00011669
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
Synonym citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
PubChem CID 311
ChEBI CHEBI:30769
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O

Specifications

Melting Point 153°C to 154°C
Beilstein 782061
Formula Weight 192.12
Density 1.665
Quantity 500 g
Packaging Plastic bottle
Physical Form Crystalline Powder
Specific Gravity 1.665
Chemical Name or Material Citric Acid Anhydrous
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.