missing translation for 'onlineSavingsMsg'
Learn More

Citric acid, Anhydrous, 99.5 to 100.5% (Dry Basis)

Product Code. 15885108 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15885108 500 g Plastic bottle 500g
15835268 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15885108 Supplier Thermo Scientific Supplier No. J13729.36
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 77-92-9 | C6H8O7 | 192.123 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-92-9
Molecular Formula C6H8O7
Molecular Weight (g/mol) 192.123
MDL Number MFCD00011669
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
Synonym citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
PubChem CID 311
ChEBI CHEBI:30769
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O

Specifications

Melting Point 153°C to 154°C
Density 1.665
Quantity 500 g
Packaging Plastic bottle
Physical Form Crystalline Powder
Specific Gravity 1.665
Beilstein 782061
Formula Weight 192.12
Chemical Name or Material Citric Acid Anhydrous

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.