missing translation for 'onlineSavingsMsg'
Learn More

Citric acid, diammonium salt, 98+%, ACS reagent

Product Code. 12615097 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12615097 500 g 500g
12605097 100 g 100g
12633620 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12615097 Supplier Thermo Scientific Acros Supplier No. 447685000
  • ONLINE EXCLUSIVE PRICE
    126.00 €
    118.00 € / 500g
    Save 8.00 €
    6% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3012-65-5 | C6H14N2O7 | 226.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3012-65-5
Molecular Formula C6H14N2O7
Molecular Weight (g/mol) 226.19
MDL Number MFCD00013068
InChI Key YXVFQADLFFNVDS-UHFFFAOYSA-N
Synonym ammonium citrate dibasic, ammonium hydrogencitrate, diammonium hydrogen citrate, citric acid diammonium salt, citric acid ammonium salt, di-ammonium hydrogen citrate, citro diamine, citric acid diamine, ammonium citrate, dibasic, acs, ammonium citrate dibasic, puriss
PubChem CID 13710713
ChEBI CHEBI:63076
IUPAC Name azane;2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES [NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O

Specifications

Density 1.5000g/mL
Ignition Residue 0.01% max
Packaging Plastic bottle
Quantity 500 g
Merck Index 15509
Specific Gravity 1.5
Solubility Information Solubility in water: soluble
Formula Weight 226.19
Grade ACS Reagent
Insoluble Matter 0.005% max
Chemical Name or Material Citric acid, diammonium salt
Show More Show Less
compliance-icons
Product Identifier
  • Citric acid, diammonium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.