missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid Electrophoresis Grade, 1kg, MP Biomedicals

Product Code. 11441562 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441562 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11441562 Supplier MP Biomedicals Supplier No. 04800681
 Restricted Item
Estimated Shipment: 08-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 77-92-9
Molecular Formula C6H8O7
MDL Number MFCD00011669
Synonym citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
Molecular Weight (g/mol) 192.12
PubChem CID 311
ChEBI CHEBI:30769
Quantity 1 kg
Chemical Name or Material Citric Acid Electrophoresis Grade
Show More Show Less
compliance-icons
Product Identifier
  • Citric Acid Anhydrous, electrophoresis grade
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: citric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.