missing translation for 'onlineSavingsMsg'
Learn More

Citric acid monohydrate, BAKER ANALYZED™ ACS, J.T. Baker™

Product Code. 10335770 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10335770 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10335770 Supplier JT Baker Supplier No. 0088.1000
  • ONLINE EXCLUSIVE PRICE
    97.98 €
    89.64 € / 1kg
    Save 8.34 €
    8% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Meets ACS Specifications

Specifications

CAS 5949-29-1
Melting Point 135°C to 152°C
Assay Percent Range 99.0 to 102.0%
Molecular Formula C6H10O8
MDL Number MFCD00149972
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
SMILES O.OC(=O)CC(O)(CC(O)=O)C(O)=O
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
Molecular Weight (g/mol) 210.14
Density 1.552 g/cm³ (20°C)
Quantity 1 kg
Flash Point 173.9°C
Packaging Plastic bottle for solids
Residue after Ignition max. 0.02%
Show More Show Less
compliance-icons
Product Identifier
  • Citric Acid Monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Citric acid, monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.