missing translation for 'onlineSavingsMsg'
Learn More

Citric acid monohydrate, Solstice

Product Code. 16655813
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
Unit Size:
1kg
2.50kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16655813 1 kg 1kg
16685813 500 g 500g
16665813 2.5 kg 2.50kg
16695813 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16655813 Supplier Fluka Supplier No. 33114H1KG
  • ONLINE EXCLUSIVE PRICE
    76.00 €
    68.78 € / 1kg
    Save 7.22 €
    9% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, 99.5-102%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5949-29-1
PubChem CID 22230
ChEBI CHEBI:31404

Specifications

Appearance Complying
Percent Purity 99.5 to 102.0 %
Grade ACS
Quantity 1 kg
Packaging Plastic Bottle
Physical Form Crystalline
compliance-icons
Product Identifier
  • Citric acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Citric acid monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.