missing translation for 'onlineSavingsMsg'
Learn More

Citric acid monohydrate, Solstice

Product Code. 16685813
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
Unit Size:
1kg
2.50kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16685813 500 g 500g
16655813 1 kg 1kg
16665813 2.5 kg 2.50kg
16695813 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16685813 Supplier Fluka Supplier No. 33114H500G
  • ONLINE EXCLUSIVE PRICE
    45.00 €
    39.15 € / 500g
    Save 5.85 €
    13% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, 99.5-102%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5949-29-1
Molecular Formula C6H10O8
Molecular Weight (g/mol) 210.14
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
PubChem CID 22230
ChEBI CHEBI:31404
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
SMILES O.OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

Appearance Complying
Percent Purity 99.5 to 102.0 %
Grade ACS
Quantity 500 g
Packaging Plastic Bottle
Physical Form Crystalline
compliance-icons
Product Identifier
  • Citric acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Citric acid monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.