missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid Monohydrate, Pharmpur™, Scharlab

Product Code. 11359933
Change view
Click to view available options
Quantity:
5 kg
Unit Size:
5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11359933 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11359933 Supplier Scharlab Supplier No. AC0720005P
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in analytical chemistry as a laboratory reagent and in buffer solutions.

  • Synonyms: 2-Hydroxy-1,2,3-propanetricarboxylic acid monohydrate, b-Hydroxytricarballylic acid monohydrate
  • Molecular Formula: C6H8O7 • H20
  • Molar Mass: 210.14g/mol
  • CAS: 5949-29-1
  • EIN ECS No.: 201-069-1
  • Oxalic acid (C2H2O4): max. 0.036%
  • Readily Carbonizable Substances: Passes test

Specifications

CAS 5949-29-1
Melting Point 135°C to 152°C
Boiling Point 135°C to 152°C (decomposition)
Molecular Formula C6H10O8
Synonym citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp
Solubility Information Very soluble in water (20°C)
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
Molecular Weight (g/mol) 210.138
PubChem CID 22230
ChEBI CHEBI:31404
Formula Weight 210.14g/mol
Percent Purity 99.5 to 100.5% (acidimetric, on dried sample)
Grade Pharmpur
Quantity 5 kg
Ignition Point 345°C
Residue after Ignition 0.05% max
Vapor Pressure <0.1hPa (at 20°C)
Show More Show Less
compliance-icons
Product Identifier
  • Citric acid monohydrate, extra pure, Pharmpur, Ph Eur, BP, USP
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.