missing translation for 'onlineSavingsMsg'
Learn More

Citric acid monohydrate, technical, crystalline

Product Code. 13238003 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 kg
Packaging:
Plastic Drum
Unit Size:
5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13238003 5 kg Plastic Drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13238003 Supplier Thermo Scientific Acros Supplier No. 444370050
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5949-29-1 | C6H8O7·H2O | 210.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5949-29-1
Melting Point 135.0°C to 152.0°C
Flash Point 174°C
Packaging Plastic Drum
Molecular Formula C6H8O7·H2O
Linear Formula HO2CCH2C(OH)(CO2H)CH2CO2H·H2O
Quantity 5 kg
Merck Index 15,2325
Synonym citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp
Solubility Information Solubility in water: 676g/L (25°C). Other solubilities: 2.17g/100g ether - 0.007g/100g chloroform, 15.43g/100g amyl alcohol - 5.98g/100g amyl acetate, 5.28g/100g ethyl acetate - 197g/100g methanol, 62.8g/100g propanol
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
Molecular Weight (g/mol) 210.15
PubChem CID 22230
ChEBI CHEBI:31404
Formula Weight 210.15
Grade Technical
Chemical Name or Material Citric Acid Monohydrate
Show More Show Less
compliance-icons
Product Identifier
  • Citric acid monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.