missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid Monohydrate, TraceSELECT™, ≥99.9998% (Metals basis), Solstice

Product Code. 15687510
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15687510 100 g Poly bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15687510 Supplier Honeywell-Fluka Supplier No. 94068100G
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
This item is not returnable. View return policy

TraceSELECT™,≥99.9998% (metals basis)

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions
TRUSTED_SUSTAINABILITY

Specifications

pH 1.8 (at 50.00g/L 20°C)
CAS 5949-29-1
Melting Point ∼135°C to 152°C
Color White
Molecular Formula C6H10O8
Linear Formula HOC(COOH)(CH2COOH)2 · H2O
MDL Number MFCD00149972
UN Number NONH for all modes of transport
Beilstein 4018641
Synonym citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
Molecular Weight (g/mol) 210.138
PubChem CID 22230
ChEBI CHEBI:31404
Formula Weight 210.14g/mol
Percent Purity ≥99.9998% (metals basis)
Grade TraceSELECT
Density ∽1.54g/cu.cm at 20°C
Quantity 100 g
Flash Point 173°C (343.4°F)
Odor Odorless
Packaging Poly bottle
Physical Form Crystalline
Chemical Name or Material Citric acid monohydrate
Show More Show Less
compliance-icons
Product Identifier
  • Citric acid monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Citric acid monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.