missing translation for 'onlineSavingsMsg'
Learn More

Citric acid, monosodium salt, 99%, for analysis, anhydrous

Product Code. 13448618 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13448618 1 kg Plastic bottle 1kg
13498678 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13448618 Supplier Thermo Scientific Acros Supplier No. 447690010
  • ONLINE EXCLUSIVE PRICE
    98.20 €
    95.30 € / 1kg
    Save 2.90 €
    2% Off
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18996-35-5 | C6H7NaO7 | 214.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18996-35-5
Molecular Formula C6H7NaO7
Molecular Weight (g/mol) 214.11
InChI Key SLWOXBQHKZPUNY-UHFFFAOYSA-N
Synonym bicitra, pneucid, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, sodium salt, sodium dihydrogen citrate anhydrous, citric acid, sodium salt, citric acid sodium, sodium 2-hydroxy-1,2,3-propanetricarboxylate, 2-hydroxypropane-1,2,3-tricarboxylic acid; sodium, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, sodium salt 1:?
PubChem CID 131675399
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;molecular hydrogen;sodium
SMILES [HH].C(C(=O)O)C(CC(=O)O)(C(=O)O)O.[Na]

Specifications

Melting Point 209.0°C to 215.0°C
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula HO2CCH2C(OH)(CO2H)CH2CO2Na
Quantity 1 kg
Solubility Information Clear colorless
Formula Weight 214.11
Percent Purity 99%
Grade Analysis
Chemical Name or Material Citric acid, monosodium salt
compliance-icons
Product Identifier
  • Citric acid, monosodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Citric acid, monosodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.