missing translation for 'onlineSavingsMsg'
Learn More

Citric acid, trisodium salt dihydrate, ACS reagent

Product Code. 11935071 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11935071 5 kg Plastic bottle 5kg
11945071 250 g Plastic bottle 250g
11925071 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11935071 Supplier Thermo Scientific Acros Supplier No. 446330050
  • ONLINE EXCLUSIVE PRICE
    289.00 €
    280.00 € / 5kg
    Save 9.00 €
    3% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6132-04-3 | C6H5Na3O7·2H2O | 294.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4-3-6132
Molecular Formula C6H5Na3O7·2H2O
Molecular Weight (g/mol) 294.09
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
Synonym trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
PubChem CID 71474
ChEBI CHEBI:32142
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]

Specifications

Melting Point >300.0°C
Packaging Plastic bottle
Linear Formula NaO2CCH2C(OH)(CO2Na)CH2CO2Na·2H2O
Quantity 5 kg
Merck Index 15,8737
Solubility Information Solubility in water: 720g/L (25°C). Other solubilities: insoluble in alcohol
Formula Weight 294.09
Grade ACS Reagent
Insoluble Matter 0.005% max
Chemical Name or Material Citric acid, trisodium salt dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.