missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid U.S.P. Granular, Macron Fine Chemicals™

Product Code. 15557854
Change view
Click to view available options
Quantity:
2.5 kg
Unit Size:
2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15557854 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15557854 Supplier Avantor Macron Supplier No. 061606
 Restricted Item
Estimated Shipment: 18-06-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 5949-29-1
Quantity 2.5 kg
Packaging Polyethylene
Physical Form Granular
Molecular Formula C6H10O8
Synonym citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
Molecular Weight (g/mol) 210.138
PubChem CID 22230
ChEBI CHEBI:31404
Formula Weight 210.14
Grade USP
Chemical Name or Material Citric Acid, Monohydrate
Show More Show Less
compliance-icons
Product Identifier
  • Citric Acid Monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Citric acid, monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.