missing translation for 'onlineSavingsMsg'
Learn More

Citronellic acid, 94%

Product Code. 11429907
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11429907

Brand: Thermo Scientific Alfa Aesar B22862.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 502-47-6 | C10H18O2 | 170.25 g/mol

Citronellic acid was used in synthesis of stereoisomers of 3,11- dimethylnonacosan-2-one and 29-hydroxy-3,11-dimethylnonacosan-2-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Citronellic acid was used in synthesis of stereoisomers of 3,11- dimethylnonacosan-2-one and 29-hydroxy-3,11-dimethylnonacosan-2-one.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 502-47-6
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.25
MDL Number MFCD00002728
InChI Key GJWSUKYXUMVMGX-UHFFFAOYNA-N
Synonym citronellic acid, 3,7-dimethyl-6-octenoic acid, rhodinolic acid, citronellate, rhodinic acid, 6-octenoic acid, 3,7-dimethyl, fema no. 3142, citronellicacid, callitrol, acmc-20apjr
PubChem CID 10402
ChEBI CHEBI:80507
IUPAC Name 3,7-dimethyloct-6-enoic acid
SMILES CC(CCC=C(C)C)CC(O)=O

Specifications

Density 0.923
Boiling Point 121°C to 122°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4535
Quantity 25 g
UN Number UN2810
Solubility Information Insoluble in water.
Formula Weight 170.25
Percent Purity 94%
Chemical Name or Material Citronellic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.